4-Methoxybenzonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
A0493-25G
A0493-5G
TCIAA0493-25GEA
2000
SEK
TCIAA0493-25G
TCIAA0493-5G
4-Methoxybenzonitrile ≥98.0%
4-Methoxybenzonitrile
Formel:
C₈H₇NO MW: 133,15 g/mol Kokpunkt: 137°C Smältpunkt: 57…60 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00001818 CAS nummer: 874-90-8 EINECS: 212-871-6 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...