3-Methoxybenzonitrile ≥95.0%

Leverantör: TCI EUROPE
Fara

M1291-25G M1291-5G
TCIAM1291-25GEA 975 SEK
TCIAM1291-25G TCIAM1291-5G
3-Methoxybenzonitrile ≥95.0%
3-Methoxybenzonitrile
Formel: C₈H₇NO
MW: 133,15 g/mol
Kokpunkt: 140°C
Smältpunkt: 23 °C
Densitet: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00001801
CAS nummer: 1527-89-5
EINECS: 216-201-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=324ef8f3f6c0ce6254e5a4976507d1265058c423&vid=a30288feb86edc11a7e8e6930b7ec89320b4b4b9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR