4-Nitrobenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

N0172-25G N0172-250G
TCIAN0172-25GEA 590 SEK
TCIAN0172-25G TCIAN0172-250G
4-Nitrobenzonitrile ≥98.0%
4-Nitrobenzonitrile
Formel: C₇H₄N₂O₂
MW: 148,12 g/mol
Smältpunkt: 145…147 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007279
CAS nummer: 619-72-7
EINECS: 210-610-0
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95cdae93ed7ef073b5f8458372764a2001cc439f&vid=df38d507067f0c104be91a49851deb286c0931ac&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR