4-Phenoxybenzaldehyde ≥98.0%
Leverantör: TCI
Synonyms:
p-Phenoxybenzenecarbaldehyde, 4-Phenoxybenzenecarbaldehyde
P1333-25G
P1333-5G
TCIAP1333-25GEA
2500
SEK
TCIAP1333-25G
TCIAP1333-5G
4-Phenoxybenzaldehyde ≥98.0%
4-Phenoxybenzaldehyde
Formel:
C₁₃H₁₀O₂ Kokpunkt: 320 °C (1013 hPa) Densitet: 1,1522 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00003383 CAS nummer: 67-36-7 EINECS: 200-650-7 |
|
Specifikation testresultat
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...