2,5-Dihydroxybenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D0565-1G
D0565-5G
TCIAD0565-1GEA
245
SEK
TCIAD0565-1G
TCIAD0565-5G
2,5-Dihydroxybenzaldehyde ≥98.0%
2,5-Dihydroxybenzaldehyde
Formel:
C₇H₆O₃ MW: 138,12 g/mol Kokpunkt: 60 - 65 °C (12 -rr) Smältpunkt: 99 °C Densitet: 0,859 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00003333 CAS nummer: 1194-98-5 EINECS: 214-789-6 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...