2,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
D0564-25G D0564-500G D0564-100G
TCIAD0564-25GEA 685 SEK
TCIAD0564-25G TCIAD0564-500G TCIAD0564-100G
2,4-Dihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
2,4-Dihydroxybenzaldehyde
Formel: C₇H₆O₃
MW: 138,12 g/mol
Kokpunkt: 228°C
Smältpunkt: 132…133 °C
Densitet: 1,448 g/cm³ (-93 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011686
CAS nummer: 95-01-2
EINECS: 202-383-1
Merck Index: 14,08156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7083b8048e2c2f8e2e5dbcac079a80dd7d1ec8fa&vid=7850e7cf39d9197c204bdfea972cc5628b67e01d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 139.0 deg-C
Solubility in Methanol almost transparency