4-Hydroxibensaldehyd 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

H0198-25G H0198-500G H0198-100G
TCIAH0198-25GEA 245 SEK
TCIAH0198-25G TCIAH0198-500G TCIAH0198-100G
4-Hydroxibensaldehyd 98,0 GC_ANALYSMETOD
4-Hydroxibensaldehyd
Formel: C₇H₆O₂
MW: 122.12 g/mol
Kokpunkt: 310
Smältpunkt: 116
Flampunkt: 174
Storage Temperature: Omgivande
MDL Number: MFCD00006939
CAS nummer: 123-08-0
EINECS: 204-599-1
Merck Index: 14,04811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b9db03c9466f34fa7d53422acb97dd8475a3245&vid=c7656203f09518b5537188ef468b1c75b8cdec60&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR