3-Hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

H0197-25G H0197-500G H0197-100G
TCIAH0197-25GEA 630 SEK
TCIAH0197-25G TCIAH0197-500G TCIAH0197-100G
3-Hydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3-Hydroxybenzaldehyde
Formel: C₇H₆O₂
MW: 122,12 g/mol
Kokpunkt: 168°C
Smältpunkt: 99…102 °C
Densitet: 1,1299 g/cm³ (114 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003368
CAS nummer: 100-83-4
EINECS: 202-892-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fadca81c1ddd2d1ba6c393feec799617e8fe2fce&vid=40a421125ff8a3caa135769cb9bda83706de0b56&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 102.0 to 105.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR