Salicylaldehyd ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
2-Hydroxybenzaldehyde
S0275-25G
S0275-500G
TCIAS0275-25GEA
430
SEK
TCIAS0275-25G
TCIAS0275-500G
Salicylaldehyd ≥98.0% (by GC, titration analysis)
Salicylaldehyd
Formel:
C₇H₆O₂ MW: 122.12 g/mol Kokpunkt: 196°C Smältpunkt: -7 Flampunkt: 90°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003317 CAS nummer: 90-02-8 EINECS: 201-961-0 Merck Index: 14,08326 |
|
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...