Salicylaldehyd ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Hydroxybenzaldehyde
S0004-25G
S0004-500G
S0004-100G
TCIAS0004-25GEA
215
SEK
TCIAS0004-25G
TCIAS0004-500G
TCIAS0004-100G
Salicylaldehyd ≥98.0%
Salicylaldehyd
Formel:
C₇H₆O₂ MW: 122,12 g/mol Kokpunkt: 196°C Smältpunkt: –7 °C Densitet: 1,17 g/cm³ (20 °C) Flampunkt: 90°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003317 CAS nummer: 90-02-8 EINECS: 201-961-0 Merck Index: 14,08326 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Density (20deg.C) | 1.1630 to 1.1680 g/mL |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...