3,4,5-Trimethoxybenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
T0059-25G
T0059-100G
TCIAT0059-25GEA
285
SEK
TCIAT0059-25G
TCIAT0059-100G
3,4,5-Trimethoxybenzaldehyde ≥98.0%
3,4,5-Trimethoxybenzaldehyde
Formel:
C₁₀H₁₂O₄ MW: 196,2 g/mol Kokpunkt: 165°C Smältpunkt: 72…75 °C Densitet: 1,367 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00003364 CAS nummer: 86-81-7 EINECS: 201-701-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 78.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...