2,4,6-Trimethoxybenzaldehyde ≥98.0%
Leverantör: TCI
Varning
T2651-25G T2651-5G
TCIAT2651-25GEA 1500 SEK
TCIAT2651-25G TCIAT2651-5G
2,4,6-Trimethoxybenzaldehyde ≥98.0%
2,4,6-Trimethoxybenzaldehyde
Formel: C₁₀H₁₂O₄
MW: 196.2 g/mol
Kokpunkt: 291
Smältpunkt: 120
Storage Temperature: Omgivande
MDL Number: MFCD00003313
CAS nummer: 830-79-5
EINECS: 212-598-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=304da6e159a3cfdf5e31b115ecab89336fde99f9&vid=b7165d8b4f5a09cda8e3cfd168da973f1b2ab731&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C