Veratraldehyde ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3,4-Dimethoxybenzaldehyde
D0507-25G D0507-500G D0507-100G
TCIAD0507-25GEA 215 SEK
TCIAD0507-25G TCIAD0507-500G TCIAD0507-100G
Veratraldehyde ≥98.0%
Veratraldehyde
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokpunkt: 281 °C (1013 hPa)
Smältpunkt: 40…43 °C
Flampunkt: ≥ 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003363
CAS nummer: 120-14-9
EINECS: 204-373-2
Merck Index: 14,09951
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94e7b5794c81ff0d70bf6056cb3ceefd901bc40c&vid=288e461417391faf1a9b591f620b8d1d2c655790&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C
Solubility in Methanol almost transparency