4-tert-Butoxybenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4-(tert-Butoxy)benzaldehyde
B3312-25G B3312-5G
TCIAB3312-25GEA 1875 SEK
TCIAB3312-25G TCIAB3312-5G
4-tert-Butoxybenzaldehyde ≥98.0%
4-tert-Butoxybenzaldehyde
Formel: C₁₁H₁₄O₂
MW: 178,23 g/mol
Flampunkt: 104°C
Storage Temperature: Omgivande
MDL Number: MFCD00040800
CAS nummer: 57699-45-3
EINECS: 260-909-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1779a4d8f9fffbb8ca8995abd1806db7fe00a190&vid=1779a4d8f9fffbb8ca8995abd1806db7fe00a190&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %