o-Anisaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde
A0479-25G
A0479-500G
A0479-100G
TCIAA0479-25GEA
215
SEK
TCIAA0479-25G
TCIAA0479-500G
TCIAA0479-100G
o-Anisaldehyde ≥98.0%
o-Anisaldehyde
Formel:
C₈H₈O₂ MW: 136,15 g/mol Kokpunkt: 243°C Smältpunkt: 34…37 °C Densitet: 1,127 g/cm³ (25 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00003308 CAS nummer: 135-02-4 EINECS: 205-171-7 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...