4-Hydroxy-3-nitrobenzaldehyde ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
H0924-25G
H0924-5G
TCIAH0924-25GEA
1400
SEK
TCIAH0924-25G
TCIAH0924-5G
4-Hydroxy-3-nitrobenzaldehyde ≥97.0%
4-Hydroxy-3-nitrobenzaldehyde
Formel:
C₇H₅NO₄ MW: 167,12 g/mol Smältpunkt: 146…148 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007117 CAS nummer: 3011-34-5 EINECS: 221-141-6 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 141.0 to 145.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...