5-Hydroxy-2-nitrobenzaldehyde ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

H1039-25G H1039-5G
TCIAH1039-25GEA 3450 SEK
TCIAH1039-25G TCIAH1039-5G
5-Hydroxy-2-nitrobenzaldehyde ≥98.0% (by GC, titration analysis)
5-Hydroxy-2-nitrobenzaldehyde
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007332
CAS nummer: 42454-06-8
EINECS: 255-832-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef3601dab37aa8144f5f1104bfc5f3901d76e60f&vid=e7a68b7f3781f05b42b78e7254d95052de072fb5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR