2-Nitrobenzaldehyd ≥99.0%

Leverantör: TCI
Varning

N0130-25G N0130-500G N0130-100G
TCIAN0130-25GEA 645 SEK
TCIAN0130-25G TCIAN0130-500G TCIAN0130-100G
2-Nitrobenzaldehyd ≥99.0%
2-Nitrobenzaldehyde
Formel: C₇H₅NO₃
MW: 151.12 g/mol
Kokpunkt: 236°C
Storage Temperature: Omgivande
MDL Number: MFCD00007132
CAS nummer: 552-89-6
EINECS: 209-025-3
Merck Index: 14,06586
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a49d25fec34193c46716fd8eef9f9f7934819c56&vid=706622d1034c6cf2ec5fb1bb3b5f4e54367172b1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 41.0 to 46.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR