2,4,6-Tribromophenol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T0349-25G T0349-500G
TCIAT0349-25GEA 215 SEK
TCIAT0349-25G TCIAT0349-500G
2,4,6-Tribromophenol ≥98.0% (by GC, titration analysis)
2,4,6-Tribromophenol
Formel: C₆H₃Br₃O
MW: 330,8 g/mol
Kokpunkt: 290 °C (1013 hPa)
Smältpunkt: 91…92 °C
Densitet: 2,55 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002150
CAS nummer: 118-79-6
EINECS: 204-278-6
UN: 3077
ADR: 9,III
Merck Index: 14,09613
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=559a258f35aeaf513a6a443ed7d52021c616f4d8&vid=183a111268b48c908f7a6c9a84c44ede0e8e5104&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 92.0 to 96.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR