2,5-Dibromo-1,4-dimethoxybenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene

D3966-25G D3966-5G
TCIAD3966-25GEA 2350 SEK
TCIAD3966-25G TCIAD3966-5G
2,5-Dibromo-1,4-dimethoxybenzene ≥98.0%
2,5-Dibromo-1,4-dimethoxybenzene
Formel: C₈H₈Br₂O₂
MW: 295,96 g/mol
MDL Number: MFCD00025694
CAS nummer: 2674-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52e7fe12312132c7af1a91e09b0762758ac7a2a&vid=22d3f61d316bfa4b769181c01b1e6dda0741613e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR