4-Bromoanisole ≥97.0%

Leverantör: TCI EUROPE

B0547-500G B0547-25G B0547-100G
TCIAB0547-500GEA 815 SEK
TCIAB0547-500G TCIAB0547-25G TCIAB0547-100G
4-Bromoanisole ≥97.0%
4-Bromoanisole
Formel: C₇H₇BrO
MW: 187,04 g/mol
Kokpunkt: 215°C
Smältpunkt: 10 °C
Densitet: 1,457 g/cm³ (20 °C)
Flampunkt: 94°C
Storage Temperature: Omgivande
MDL Number: MFCD00000097
CAS nummer: 104-92-7
EINECS: 203-252-1
Merck Index: 14,01428
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859125b799f9f3e651e9034406cca25a7e66b0e3&vid=d8869bf90f325b7c5cd0164e9799fb17f86d2306&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR