2-Bromoanisole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-Bromo-2-methoxybenzene
B0546-25G
B0546-500G
B0546-100G
TCIAB0546-25GEA
310
SEK
TCIAB0546-25G
TCIAB0546-500G
TCIAB0546-100G
2-Bromoanisole ≥98.0%
2-Bromoanisole
Formel:
C₇H₇BrO MW: 187,04 g/mol Kokpunkt: 223°C Densitet: 1,5 g/cm³ (20 °C) Flampunkt: 101°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000064 CAS nummer: 578-57-4 EINECS: 209-425-8 ADR: 9,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...