4-Bromo-3-nitroanisole ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Bromo-4-methoxy-2-nitrobenzene
B3420-25G B3420-5G
TCIAB3420-25GEA 1600 SEK
TCIAB3420-25G TCIAB3420-5G
4-Bromo-3-nitroanisole ≥96.0%
4-Bromo-3-nitroanisole
Formel: C₇H₆BrNO₃
MW: 232,03 g/mol
Kokpunkt: 154°C
Storage Temperature: Kylskåp
MDL Number: MFCD00051511
CAS nummer: 5344-78-5
EINECS: 226-290-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=813eb2ee2930e5a8c5920ba887fbb82dd8769c10&vid=362b1402796bfc3241f8dcc2803a2570c4f333f7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 96.0 %