1-Bromo-2-nitrobenzene ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Bromonitrobenzene
B0622-25G
B0622-500G
B0622-100G
TCIAB0622-25GEA
510
SEK
TCIAB0622-25G
TCIAB0622-500G
TCIAB0622-100G
1-Bromo-2-nitrobenzene ≥99.0%
1-Bromo-2-nitrobenzene
Formel:
C₆H₄BrNO₂ MW: 202,01 g/mol Kokpunkt: 264°C Smältpunkt: 39…42 °C Densitet: 1,62 g/cm³ (20 °C) Flampunkt: 112 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007045 CAS nummer: 577-19-5 EINECS: 209-409-0 UN: 3459 ADR: 6.1,III |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 40.0 to 43.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...