2-Bromoaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
B0541-25G B0541-250G
TCIAB0541-25GEA 390 SEK
TCIAB0541-25G TCIAB0541-250G
2-Bromoaniline ≥98.0% (by GC, titration analysis)
2-Bromoaniline
Formel: C₆H₆BrN
MW: 172,02 g/mol
Kokpunkt: 229 °C (1013 hPa)
Smältpunkt: 27…31 °C
Densitet: 1,86 g/cm³ (20 °C)
Flampunkt: ≥ 110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007632
CAS nummer: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2501016c67dcd17f771dc5f346f4abce627d16cb&vid=47c6bdc21f5690f0ad625f5d6f106cb50f054429&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %