2,4-Dibromoaniline ≥98.0%

Leverantör: TCI
Fara

D1153-10G D1153-25G
TCIAD1153-10GEA 405 SEK
TCIAD1153-10G TCIAD1153-25G
2,4-Dibromoaniline ≥98.0%
2,4-Dibromoaniline
Formel: C₆H₅Br₂N
Kokpunkt: 156°C
Smältpunkt: 80…82 °C
Densitet: 2,26 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007633
CAS nummer: 615-57-6
EINECS: 210-434-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31824ce5012d5154f049cdae5779997716ec4139&vid=2fd3e5bc1819cded3b48807d487fa0e7645180e5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR