2,4,6-Tribromoaniline ≥99.0%

Leverantör: TCI
Varning

T1088-25G T1088-500G
TCIAT1088-25GEA 350 SEK
TCIAT1088-25G TCIAT1088-500G
2,4,6-Tribromoaniline ≥99.0%
2,4,6-Tribromoaniline
Formel: C₆H₄Br₃N
Kokpunkt: 300 °C (1013 hPa)
Smältpunkt: 118 °C
Densitet: 2,578 g/cm³ (20 °C)
MDL Number: MFCD00007634
CAS nummer: 147-82-0
EINECS: 205-700-1
ADR: 6.1,II
Merck Index: 14,09610
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dfe3735f02ed1c9c12278307941b4380d8cccfd&vid=429fd4cf9a90b9cb9b277df7a10e50c855bd35d2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 119.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR