1-Bromo-2-fluoro-4-iodobenzene ≥97.0%
Leverantör: TCI
|
Varning
|
B3017-25G
B3017-5G
TCIAB3017-25GEA
1925
SEK
TCIAB3017-25G
TCIAB3017-5G
1-Bromo-2-fluoro-4-iodobenzene ≥97.0%
1-Bromo-2-fluoro-4-iodobenzene
Formel:
C₆H₃BrFI Kokpunkt: 112°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00042577 CAS nummer: 136434-77-0 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...