2-Bromo-5-fluorobenzonitrile ≥97.0%
Leverantör: TCI EUROPE
Fara
B3010-25G B3010-5G
TCIAB3010-25GEA 1825 SEK
TCIAB3010-25G TCIAB3010-5G
2-Bromo-5-fluorobenzonitrile ≥97.0%
2-Bromo-5-fluorobenzonitrile
Formel: C₇H₃BrFN
MW: 200,01 g/mol
Kokpunkt: 255 °C (1013 hPa)
Smältpunkt: 89…91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00142875
CAS nummer: 57381-39-2
EINECS: 260-711-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d7c0e0751a6b1bd6708358246b020bd72c12827&vid=ab90bfb197c907f8cf41623d5de9139f10d57898&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 88.0 to 92.0 deg-C