2-Amino-3-benzyloxypyridine ≥98.0%

Leverantör: TCI
Varning

A2080-25G A2080-5G
TCIAA2080-25GEA 1250 SEK
TCIAA2080-25G TCIAA2080-5G
2-Amino-3-benzyloxypyridine ≥98.0%
2-Amino-3-benzyloxypyridine
Formel: C₁₂H₁₂N₂O
MDL Number: MFCD00006316
CAS nummer: 24016-03-3
EINECS: 245-983-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7b62d99758efc68257c6a3d1f1b9e6a3a06db42&vid=e7b62d99758efc68257c6a3d1f1b9e6a3a06db42&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR