2-(2,4-Dinitrobenzyl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

D1095-1G D1095-5G
TCIAD1095-1GEA 245 SEK
TCIAD1095-1G TCIAD1095-5G
2-(2,4-Dinitrobenzyl)pyridine ≥98.0%
2-(2,4-Dinitrobenzyl)pyridine
Formel: C₁₂H₉N₃O₄
MW: 259,22 g/mol
MDL Number: MFCD00014632
CAS nummer: 1151-97-9
EINECS: 214-567-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d39dc4dc20b79d8cf615bfd3049f683c4c993a&vid=60d39dc4dc20b79d8cf615bfd3049f683c4c993a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR