3-Nitrophenylhydrazinium chloride ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Nitrophenylhydrazine hydrochloride, 1-(3-Nitrophenyl)hydrazine hydrochloride, (3-Nitrophenyl)hydrazine hydrochloride

N0232-25G N0232-5G
TCIAN0232-25GEA 1700 SEK
TCIAN0232-25G TCIAN0232-5G
3-Nitrophenylhydrazinium chloride ≥98.0%
3-Nitrophenylhydrazinium chloride
Formel: C₆H₈ClN₃O₂
MW: 189,6 g/mol
MDL Number: MFCD00012939
CAS nummer: 636-95-3
EINECS: 211-270-6
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55c2ee8510870e3534cb6957af95796dd3870735&vid=55c2ee8510870e3534cb6957af95796dd3870735&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Solubility in hot Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR