3-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride

C1477-25G C1477-5G
TCIAC1477-25GEA 925 SEK
TCIAC1477-25G TCIAC1477-5G
3-Chlorophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
3-Chlorophenylhydrazinium chloride
Formel: C₆H₈Cl₂N₂
MW: 179,05 g/mol
MDL Number: MFCD00012935
CAS nummer: 2312-23-4
EINECS: 219-005-6

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR