Benzophenone hydrazone ≥98.0%

Leverantör: TCI EUROPE

B1163-25G B1163-500G
TCIAB1163-25GEA 215 SEK
TCIAB1163-25G TCIAB1163-500G
Benzophenone hydrazone ≥98.0%
Benzophenone hydrazone
Formel: C₁₃H₁₂N₂
MW: 196,25 g/mol
Kokpunkt: 230°C
MDL Number: MFCD00007624
CAS nummer: 5350-57-2
EINECS: 226-321-8
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d5050bc6288590c75ebfd40f156328573b9493&vid=e6d5050bc6288590c75ebfd40f156328573b9493&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR