Ethylammonium chloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Aminoethane hydrochloride, Ethylamine hydrochloride

E0205-25G E0205-500G
TCIAE0205-25GEA 215 SEK
TCIAE0205-25G TCIAE0205-500G
Ethylammonium chloride ≥98.0%
Ethylammonium chloride
Formel: C₂H₈ClN
MW: 81,54 g/mol
MDL Number: MFCD00012885
CAS nummer: 557-66-4
EINECS: 209-182-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fdf7b0e84a3758f05fc54857baec2db1036ade8&vid=9fdf7b0e84a3758f05fc54857baec2db1036ade8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Melting point 110.0 to 115.0 deg-C
Solubility in Water almost transparency
Water max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR