4-Fluorophenyl methyl sulphone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Fluoro-4-(methylsulphonyl)benzene, 1-Fluor-4-(methylsulphonyl)benzol
F0651-25G
F0651-5G
TCIAF0651-25GEA
1950
SEK
TCIAF0651-25G
TCIAF0651-5G
4-Fluorophenyl methyl sulphone ≥98.0%
4-Fluorophenyl methyl sulphone
Formel:
C₇H₇FO₂S MW: 174,2 g/mol |
MDL Number:
MFCD00039753 CAS nummer: 455-15-2 EINECS: 207-237-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...