Chlorodimethylvinylsilane ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Dimethylvinylchlorosilane, Vinyldimethylchlorosilane

C1208-25ML C1208-100ML
TCIAC1208-25MLEA 415 SEK
TCIAC1208-25ML TCIAC1208-100ML
Chlorodimethylvinylsilane ≥97.0%
Chlorodimethylvinylsilane
Formel: C₄H₉ClSi
MW: 120,65 g/mol
Kokpunkt: 82°C
Flampunkt: –6°C
MDL Number: MFCD00018090
CAS nummer: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fba13fcf8f1018aa048e04aee812979069b3314&vid=1fba13fcf8f1018aa048e04aee812979069b3314&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR