(Chloromethyl)dimethylchlorosilane ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
CMDMCS, Chloro(chloromethyl)dimethylsilane
C0605-25G
C0605-250G
TCIAC0605-25GEA
685
SEK
TCIAC0605-25G
TCIAC0605-250G
(Chloromethyl)dimethylchlorosilane ≥98.0%
(Chloromethyl)dimethylchlorosilane
Formel:
C₃H₈Cl₂Si MW: 143,09 g/mol Kokpunkt: 115°C Flampunkt: 22°C |
MDL Number:
MFCD00000875 CAS nummer: 1719-57-9 EINECS: 217-006-6 UN: 2985 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...