1,4-Dibromotetrafluorobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

D1493-25G D1493-5G
TCIAD1493-25GEA 3425 SEK
TCIAD1493-25G TCIAD1493-5G
1,4-Dibromotetrafluorobenzene ≥98.0%
1,4-Dibromotetrafluorobenzene
Formel: C₆Br₂F₄
MW: 307,87 g/mol
Kokpunkt: 198 °C (1013 hPa)
Smältpunkt: 78,2…78.5 °C
MDL Number: MFCD00000309
CAS nummer: 344-03-6
EINECS: 206-448-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0a16290283aceab2a43a34ed3c3571ff253ca3&vid=7df0d409f52ed59e38bde7e7b846779b0a4692ff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR