2,4-Dibromo-1-fluorobenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,3-Dibromo-4-fluorobenzene, 1-Fluoro-2,4-dibromobenzene
D3489-25G
D3489-5G
TCIAD3489-25GEA
1800
SEK
TCIAD3489-25G
TCIAD3489-5G
2,4-Dibromo-1-fluorobenzene ≥97.0%
2,4-Dibromo-1-fluorobenzene
Formel:
C₆H₃Br₂F MW: 253,9 g/mol Kokpunkt: 105°C Flampunkt: 92°C |
MDL Number:
MFCD00000283 CAS nummer: 1435-53-6 EINECS: 215-860-4 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...