4-Bromo-2,6-dichloroaniline ≥98.0%

Leverantör: TCI EUROPE
Varning

B1978-25G B1978-5G
TCIAB1978-25GEA 1825 SEK
TCIAB1978-25G TCIAB1978-5G
4-Bromo-2,6-dichloroaniline ≥98.0%
4-Bromo-2,6-dichloroaniline
Formel: C₆H₄BrCl₂N
MW: 240,91 g/mol
Kokpunkt: 290 °C (1013 hPa)
Smältpunkt: 90 °C
MDL Number: MFCD00025162
CAS nummer: 697-88-1
EINECS: 211-809-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c3076c4fb29f323258fc265576cbaef65c7e4c8&vid=8929130f8181fbe8fc71633f75bd9fc9a0850ff8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR