1-Bromo-4-chlorobenzene ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4-Bromochlorobenzene
B0571-25G B0571-500G B0571-100G
TCIAB0571-25GEA 245 SEK
TCIAB0571-25G TCIAB0571-500G TCIAB0571-100G
1-Bromo-4-chlorobenzene ≥99.0%
1-Bromo-4-chlorobenzene
Formel: C₆H₄BrCl
MW: 191,45 g/mol
Kokpunkt: 196 °C (1013 hPa)
Smältpunkt: 66…68 °C
Densitet: 1,576 g/cm³ (71 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000600
CAS nummer: 106-39-8
EINECS: 203-392-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8df08256b77ce27db9eb1d056c3f884af1b843c2&vid=c5c210fefaf3b78bce3feeec14286f01fab55475&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 65.0 to 67.0 deg-C
Solubility in Methanol almost transparency