1-Bromo-3-chlorobenzene ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Bromochlorobenzene
B0570-100G
B0570-25G
TCIAB0570-100GEA
645
SEK
TCIAB0570-100G
TCIAB0570-25G
1-Bromo-3-chlorobenzene ≥99.0%
1-Bromo-3-chlorobenzene
Formel:
C₆H₄BrCl MW: 191,45 g/mol Kokpunkt: 196°C Smältpunkt: –21,2 °C Densitet: 1,63 g/cm³ (20 °C) Flampunkt: 80°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000568 CAS nummer: 108-37-2 EINECS: 203-575-8 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...