2-Bromobenzonitrile ≥98.0%

Leverantör: TCI
Varning

B1369-10G B1369-25G
TCIAB1369-10GEA 535 SEK
TCIAB1369-10G TCIAB1369-25G
2-Bromobenzonitrile ≥98.0%
2-Bromobenzonitrile
Formel: C₇H₄BrN
Kokpunkt: 252 °C (1013 hPa)
Smältpunkt: 51…52 °C
Densitet: 1,496 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001772
CAS nummer: 2042-37-7
EINECS: 218-045-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05a95b8ad2ef230095628341ed016367539146d2&vid=75944ed1153a7e8e3f4ff355664c7ad5aa2bd95e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 56.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR