3-Bromo-4-hydroxi-5-metoxibensaldehyd 95,0 Titrimetrisk

Leverantör: TCI
Varning

Synonyms: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde

B1172-25G B1172-5G
TCIAB1172-25GEA 1050 SEK
TCIAB1172-25G TCIAB1172-5G
3-Bromo-4-hydroxi-5-metoxibensaldehyd 95,0 Titrimetrisk
3-Bromo-4-hydroxi-5-metoxibensaldehyd
Formel: C₈H₇BrO₃
MW: 231.05 g/mol
Kokpunkt: 314
MDL Number: MFCD00006940
CAS nummer: 2973-76-4
EINECS: 221-016-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d50e2bee1f5370f8ff28e3d94b9b097950dbfe5&vid=6d50e2bee1f5370f8ff28e3d94b9b097950dbfe5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 161.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR