2-Amino-3,5-dibromobenzaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

D2625-25G D2625-5G
TCIAD2625-25GEA 885 SEK
TCIAD2625-25G TCIAD2625-5G
2-Amino-3,5-dibromobenzaldehyde ≥98.0%
2-Amino-3,5-dibromobenzaldehyde
Formel: C₇H₅Br₂NO
MW: 278,93 g/mol
Kokpunkt: 331 °C (1013 hPa)
Smältpunkt: 137…137,5 °C
MDL Number: MFCD00671100
CAS nummer: 50910-55-9
EINECS: 256-841-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4cce03c5651a0e220495b08bd560cbee6fb5437&vid=77c0b691d96751f751c8781967d82334a729d321&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 135.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR