2-Bromobenzaldehyde ≥98.0%
Leverantör: TCI
Varning
B0836-25G B0836-500G B0836-100G
TCIAB0836-25GEA 335 SEK
TCIAB0836-25G TCIAB0836-500G TCIAB0836-100G
2-Bromobenzaldehyde ≥98.0%
2-Bromobenzaldehyde
Formel: C₇H₅BrO
Kokpunkt: 230°C
Smältpunkt: 21 °C
Densitet: 1,58 g/cm³ (20 °C)
Flampunkt: 95°C
Storage Temperature: Omgivande
MDL Number: MFCD00003300
CAS nummer: 6630-33-7
EINECS: 229-622-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b58b4e3b342b5db44e6e89da867a9abf14c3237&vid=502418375331fbf00d09a59d261b6b0effa48609&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 14.0 to 22.0 deg-C