1,3,5-Tribromobenzene ≥98.0%
Leverantör: TCI EUROPE
Varning
T0347-25G T0347-250G T0347-5G
TCIAT0347-25GEA 325 SEK
TCIAT0347-25G TCIAT0347-250G TCIAT0347-5G
1,3,5-Tribromobenzene ≥98.0%
1,3,5-Tribromobenzene
Formel: C₆H₃Br₃
MW: 314,8 g/mol
Kokpunkt: 271°C
Smältpunkt: 122…123 °C
Densitet: 2,637 g/cm³ (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000080
CAS nummer: 626-39-1
EINECS: 210-947-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=531bd0c820652d813956db81402b2876926fd3bd&vid=3a292f671c78068012e19d867eea0bda97660995&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 124.0 deg-C
Solubility in hot Methanol almost transparency