Methyl-4-iodo-2-methoxybenzoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate
B25529.06 B25529.14
B25529.06EA 1650 SEK
B25529.06 B25529.14
Methyl-4-iodo-2-methoxybenzoate ≥98%
Methyl-4-iodo-2-methoxybenzoate
Purity: 98+% CAS No.: 148490-97-5 Molecular Formula: C9H9IO3 Formula Weight: 292.07 Melting Point: 44-46° MDL No.: MFCD06797865
Formel: C₉H₉IO₃
MW: 292,07 g/mol
Smältpunkt: 44…46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052925
CAS nummer: 35387-93-0