1,2,4,5-Tetrabromobenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
T0031-25G
T0031-5G
TCIAT0031-25GEA
2775
SEK
TCIAT0031-25G
TCIAT0031-5G
1,2,4,5-Tetrabromobenzene ≥97.0%
1,2,4,5-Tetrabromobenzene
Formel:
C₆H₂Br₄ MW: 393,7 g/mol |
MDL Number:
MFCD00000063 CAS nummer: 636-28-2 EINECS: 211-253-3 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 178.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...